Tisk…
Masterflex® Flow Indicators and Switches for Liquids, Avantor®
Dodavatel: Avantor Fluid Handling
32880-20
MFLX32880-20EA 14280 CZK
MFLX32880-20
Masterflex® Flow Indicators and Switches for Liquids, Avantor®
Indikátory průtoku
Easily view flow through clear plastic lens.
  • Constructed with polypropylene (PP) housing
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators feature a bright orange rotor that spins to provide visual verification.
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, andMore Product Information