Tisk…
McMillan Flow High-Accuracy Stainless Steel Flowmeters and Transmitters
Dodavatel: Avantor Fluid Handling
32718-06 32718-22 32718-24 32718-26 32718-30 32718-48 32718-02 32718-07 32718-09
MFLX32718-06EA 22450 CZK
MFLX32718-06 MFLX32718-22 MFLX32718-24 MFLX32718-26 MFLX32718-30 MFLX32718-48 MFLX32718-02 MFLX32718-07 MFLX32718-09
McMillan Flow High-Accuracy Stainless Steel Flowmeters and Transmitters
Průtokoměry
Economical design with an outstanding ±1.0 % full-scale accuracy.
  • A wide range of outputs options for integration into any system
Select these flowmeters and transmitters for extremely low flow-rate applications or for higher flow rate measurements and yet maintain a full-scale accuracy of ±1% or better. All units feature a stainless steel housing and come standard with stainless steel compression fittings.
The flowmeters/transmitters can be used with a wide variety of transparent, low-viscosity (under 10 cSt) corrosive or neutral liquids and solvents.More Product Information