Tisk…
Air/N2 Gas Mass Flow Controllers with Metal bodies
Dodavatel: Avantor Fluid Handling
32660-04 32660-06 32660-08 32660-10 32660-12 32660-14 32660-16 32660-19 32660-22 32660-24 32661-00 32661-04 32661-08 32661-12 32661-16 32661-22 32665-10 32666-00
MFLX32660-04EA 26500 CZK
MFLX32660-04 MFLX32660-06 MFLX32660-08 MFLX32660-10 MFLX32660-12 MFLX32660-14 MFLX32660-16 MFLX32660-19 MFLX32660-22 MFLX32660-24 MFLX32661-00 MFLX32661-04 MFLX32661-08 MFLX32661-12 MFLX32661-16 MFLX32661-22 MFLX32665-10 MFLX32666-00
Air/N2 Gas Mass Flow Controllers with Metal bodies
Průtokové spínače
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established byMore Product Information