Tisk…
CPC® DrumQuik® PRO Drum Dispensing System Components
Dodavatel: Avantor Fluid Handling
31608-20 31608-21 31608-22 31608-25 31608-38 31608-20BLK 31608-22BLK 31608-21BLK 31608-25BLK
MFLX31608-20EA 3400 CZK
MFLX31608-20 MFLX31608-21 MFLX31608-22 MFLX31608-25 MFLX31608-38 MFLX31608-20BLK MFLX31608-22BLK MFLX31608-21BLK MFLX31608-25BLK
CPC® DrumQuik® PRO Drum Dispensing System Components
Fitinky a konektory
Closed-system design ensures the safety of your environment and employees - at all phases of chemical handling.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Indivdual components to keep your dispensing system working smoothly - or build your own
  • Easy-to-use, economical solution for chemical dispensing
  • Durable materials withstand harsh operating conditions
  • Ideal for 5-gallon jerry cans, 55-gallon drums, and IBCs
The CPC® DrumQuik® PRO drum dispensing system offers a simplified solution to your chemical dispensing needs. This systemMore Product Information