Tisk…
CPC® Metal Quick-Disconnect Fittings, NPT (M) Threaded Inserts
Dodavatel: Avantor Fluid Handling
31403-50 31403-55 31403-56 31404-60 31404-61 31403-50BLK 31403-55BLK 31403-56BLK 31404-61BLK 31404-60BLK
MFLX31403-50EA 795 CZK
MFLX31403-50 MFLX31403-55 MFLX31403-56 MFLX31404-60 MFLX31404-61 MFLX31403-50BLK MFLX31403-55BLK MFLX31403-56BLK MFLX31404-61BLK MFLX31404-60BLK
CPC® Metal Quick-Disconnect Fittings, NPT (M) Threaded Inserts
Fitinky a konektory
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from −40 toMore Product Information