Tisk…
CPC® Metal Quick-Disconnect Fittings, Hose Barb Inserts
Dodavatel: Avantor Fluid Handling
31403-40 31403-42 31403-45 31403-40BLK 31403-42BLK 31404-51BLK
MFLX31403-40EA 783 CZK
MFLX31403-40 MFLX31403-42 MFLX31403-45 MFLX31403-40BLK MFLX31403-42BLK MFLX31404-51BLK
CPC® Metal Quick-Disconnect Fittings, Hose Barb Inserts
Fitinky a konektory
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from −40 toMore Product Information