Tisk…
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
Dodavatel: Avantor Fluid Handling
31404-49 31404-69 31404-49BLK 31404-69BLK 31308-84BLK 31309-21BLK 31308-85BLK 31309-11BLK 31309-19BLK
MFLX31404-49EA 438 CZK
MFLX31404-49 MFLX31404-69 MFLX31404-49BLK MFLX31404-69BLK MFLX31308-84BLK MFLX31309-21BLK MFLX31308-85BLK MFLX31309-11BLK MFLX31309-19BLK
CPC® Metal Quick-Disconnect Fittings, Compression Inserts
Fitinky a konektory
Convenience of a quick-disconnect in pressure and vacuum applications.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Constructed of durable chrome-plated brass
  • Easy one-hand connection and disconnection
These CPC® metal quick-disconnect fittings are ideal for pressure and vacuum applications. Fittings are made of chrome-plated brass with stainless steel springs and latch. Valved fittings have an acetal shutoff valve that seals the flow path when fitting halves are disconnected. Can withstand vacuum to 250 psi, temperatures from −40 toMore Product Information