Tisk…
Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Dodavatel: Avantor Fluid Handling
40614-59 40614-67
MFLX30703-49EA 900 CZK
MFLX30703-49 MFLX30703-55 MFLX30703-56 MFLX40703-41 MFLX40614-59 MFLX40614-67 MFLX40614-51 MFLX40614-53 MFLX40614-56 MFLX40614-52 MFLX40614-55 MFLX40614-57 MFLX40614-58 MFLX40614-60 MFLX40614-61 MFLX40614-62 MFLX40614-65 MFLX40614-66 MFLX40614-68 MFLX40614-69 MFLX40614-70 MFLX40614-71 MFLX40614-72 MFLX40614-73 MFLX40614-74 MFLX40614-75 MFLX40614-76 MFLX40614-77 MFLX40614-78 MFLX40614-63
Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF), Avantor®
Fitinky a konektory
Best for low-pressure applications.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture