Tisk…
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Dodavatel: Avantor Fluid Handling
29590-80 29590-83 30548-22
MFLX30548-00EA 625 CZK
MFLX30548-00 MFLX30548-02 MFLX30548-04 MFLX30548-06 MFLX30548-08 MFLX30548-20 MFLX30548-24 MFLX30548-26 MFLX30548-28 MFLX30548-40 MFLX30548-42 MFLX30548-44 MFLX30548-46 MFLX30548-60 MFLX30548-62 MFLX30548-64 MFLX30548-66 MFLX30548-68 MFLX30548-80 MFLX30548-82 MFLX30548-84 MFLX30548-86 MFLX30548-88 MFLX00162-NJ MFLX16011-66 MFLX41001-77 MFLX29590-80 MFLX29590-83 MFLX30548-22 MFLX30548-48 MFLX00364-YQ MFLX00404-MO
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Těsnění Těsnění
Ensure a tight seal.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food,More Product Information