Tisk…
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Dodavatel: Avantor Fluid Handling
30542-09 30542-10 30542-17 30542-18 30543-05 30543-13 30543-18 30544-00 30544-05 30544-06 30544-14 30545-12 30545-14 30545-16
MFLX30542-00EA 1980 CZK
MFLX30542-00 MFLX30542-02 MFLX30542-04 MFLX30542-05 MFLX30542-06 MFLX30542-08 MFLX30542-12 MFLX30542-13 MFLX30542-14 MFLX30542-16 MFLX30543-00 MFLX30543-04 MFLX30543-08 MFLX30543-14 MFLX30543-16 MFLX30544-04 MFLX30544-12 MFLX30544-16 MFLX30545-13 MFLX30542-01 MFLX30543-12 MFLX30542-09 MFLX30542-10 MFLX30542-17 MFLX30542-18 MFLX30543-05 MFLX30543-13 MFLX30543-18 MFLX30544-00 MFLX30544-05 MFLX30544-06 MFLX30544-14 MFLX30545-12 MFLX30545-14 MFLX30545-16
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Těsnění Těsnění
Smart Gasket lets you take samples or readings from a sanitary line.
  • Measure temperature, take a sample, or dose chemicals into your process line
  • Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.