Tisk…
Masterflex® Smart Gasket® 1-Port Sanitary Gaskets, Silicone, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX30542-20EA 3730 CZK
MFLX30542-20
Masterflex® Smart Gasket® 1-Port Sanitary Gaskets, Silicone, Avantor®
Těsnění Těsnění
Easily sample fluid in critical high-purity system.
  • One internal port accepts thermocouple probe
Masterflex® Smart Gasket® is ideal when validating sterility in a high-purity pharmaceutical system. The Smart Gasket is used to obtain the critical thermal mapping information you need during the validation process. It easily installs between two sanitary standard ferrules.