Tisk…
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, PTFE, Avantor®
Dodavatel: Avantor Fluid Handling
30538-04 30538-24 30538-44 30538-46 30538-60 30538-66
MFLX30538-06EA 4300 CZK
MFLX30538-06 MFLX30538-08 MFLX30538-20 MFLX30538-26 MFLX30538-28 MFLX30538-40 MFLX30538-48 MFLX30538-64 MFLX30538-68 MFLX30538-80 MFLX30538-84 MFLX30538-86 MFLX30538-88 MFLX61030-13 MFLX30538-04 MFLX30538-24 MFLX30538-44 MFLX30538-46 MFLX30538-60 MFLX30538-66
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, PTFE, Avantor®
Těsnění Těsnění
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information