Tisk…
Hand-Held Tubing Cutter
Dodavatel: Avantor Fluid Handling
MFLX06438-90EA 1460 CZK
MFLX06438-90
Hand-Held Tubing Cutter
Sekáčky
Get clean, straight cuts everytime.
  • Cuts through tubing up to 1 1/4" OD
Steel blade coated with PTFE® resin produces clean, straight cuts through tubing up to 1-1/4" OD (cuts polyethylene tubing up to 1-1/2" OD). Optional carrying pouch and replacement blade available.