Tisk…
Masterflex® L/S® High-Performance Precision Pump Tubing, PharmaPure®, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX06435-15EA 7950 CZK
MFLX06435-15 MFLX06435-24
Masterflex® L/S® High-Performance Precision Pump Tubing, PharmaPure®, Avantor®
Hadičky Hadičky peristaltické pumpy
Ultra-low spallation protects fluid purity – ideal for sensitive or costly fluids and for filtration applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
More Product Information