Tisk…
Masterflex® L/S® Spooled Pump Tubing, C-Flex®, Avantor®
Dodavatel: Avantor Fluid Handling
MFLX06427-14EA 19400 CZK
MFLX06427-14 MFLX06427-15 MFLX06427-16 MFLX06427-17 MFLX06427-18 MFLX06427-24 MFLX06427-35 MFLX06427-36 MFLX06427-25 MFLX06427-13
Masterflex® L/S® Spooled Pump Tubing, C-Flex®, Avantor®
Hadičky Hadičky peristaltické pumpy
Combines biocompatibility of silicone with chemical resistance similar to Tygon®.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs,
More Product Information