Tisk…
Redox buffer solutions
Dodavatel: Mettler - Toledo
51350064 51350062 51350060
METT51350064EA 1100 CZK
METT51350064 METT51350062 METT51350060
Redox buffer solutions
Pufry Pufry pH
Redox buffer solutions are used for verification of all common redox sensors.
  • Temperature dependent
  • Used in quality assurance systems
A table on every bottle label indicates redox values at different temperatures and hence not used for calibration purposes.
Informace o dodávce: Supplied with a quality inspection certificate.