Tisk…
Titrators, Karl Fisher, EasyPlus™
Dodavatel: Mettler - Toledo
30060041 30060045
METN30060041EA 111680 CZK
METN30060041 METT30060045
Titrators, Karl Fisher, EasyPlus™
Titrátory
This versatile titrator is suitable for a range of determinations making it ideal for simple titrations of most sample types. The simple 'App' style user interface reduces training time while increasing work speed and simplicity. iTitrate Intelligence’s guided set-up assists in method programming to ensure samples are tested consistently, regardless of operator, and with minimal effort. Ideal for KF titrations of a wide range of samples from any industry.
  • Very small footprint
  • LongClick™ function starts routine analysis from the home screen
  • iTitrate