Tisk…

Accessories for E.Z.N.A.® MicroElute kits

Dodavatel: OMEGA BIO-TEK

MEDNACOL-02
MEDNACOL-02EA 11110 CZK
MEDNACOL-02
Accessories for E.Z.N.A.® MicroElute kits
Činidla pro nukleové kyseliny Nucleic Acid Purification Kits and Reagents

Objednat

Produktová specifikace

Environmentally Preferable
Aplikace
Target molecule
Refine target molecule
Sample type
Method / Format

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR