Tisk…
10% SDS (Sodium Dodecyl Sulfate), Corning®
Dodavatel: Corning
46-040-CI
MDTC46-040-CIEA 1720 CZK
MDTC46-040-CI
10% SDS (Sodium Dodecyl Sulfate), Corning®
Pufry Pufry pro tkáňové kultury
Sodium Dodecyl Sulfate (SDS) is an anionic detergent with applications in SDS-PAGE protein analysis and nucleic acid purification. It is a wetting agent that is effective in both acid and alkaline solutions, and is most often used as a protein and lipid solubilization reagent. SDS is also a powerful protein denaturant. This 10% solution is prepared in 18 megaohm water and is 0.2µm filtered.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM