Tisk…

Gentamicin sulphate solution 50 mg/ml in deionised water

Dodavatel: Corning

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

30-005-CR
MDTC30-005-CREA 17310 CZK
MDTC30-005-CR
Gentamicin sulphate solution 50 mg/ml in deionised water
Gentamicin sulphate

Gentamicin works as an antimicrobial agent by binding to the 30S ribosomal subunit of certain bacteria.


It is formulated to contain 50 mg/ml gentamicin in deionized water and is recommended for use in cell culture applications at 1 ml/L.


This product is effective against mycoplasma, Gram-negative and Gram-positive bacteria and is stable at 37°C for 5 days.


Balení: Shipping temperature: Ambient

Storage Temperature: Ambient
MDL Number: MFCD00270181
CAS číslo: 1405-41-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR