Tisk…
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Dodavatel: Corning
25-800-CR
MDTC25-800-CREA 1800 CZK
MDTC25-800-CR
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Doplňky a přísady médií z buněčných kultur
Animal free formulation
Contains:
1000 mg/L insulin, human recombinant
550 mg/L transferrin, human recombinant
0.67 mg/L selenious acid
Upozornení: Storage 2°C to 8°C. For research use only.