Tisk…
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 2 µm
Dodavatel: Avantor
MDKA-2-1002UEA 33010 CZK
MDKA-2-1002U MDKA-2-1546U MDKA-2-1503U MDKA-2-1502U MDKA-2-1046U MDKA-2-1003U MDKA-2-0546U MDKA-2-0503U MDKA-2-0502U
Avantor® ACE®, HPLC/UHPLC Columns, Advanced Method Development Kits, 2 µm
Kolony chromatografické
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1,7 µm and 5 µm ) and complementary phases
  • Analytical IDs in
More Product Information