Tisk…
Cut resistant gloves, Cutstar
Dodavatel: Marigold (Ansell)
2FD0EP4000 2FD0EP2000
MARG2FD0EP4000EA 0 CZK
MARG2FD0EP4000 MARG2FD0EP2000
Cut resistant gloves, Cutstar
Rukavice
These gloves are made from high performance synthetic fibres using the Techcor® Knitting Technology which ensures a fine and close knitting. The gloves feature a PVC dot coating on palm and fingers for improved grip performance.
  • Excellent protection against cutting
  • Very good resistance to abrasion
  • Glove knitted with a seamless non-stop process
  • Excellent dexterity due to fine knitting
  • Elasticated knit wrist to ensure snug fit
Ideal for all kinds of assembling work requiring dexterity, precision and protection against sharp objects.
Certifikace: EN 38More Product Information
Objednat
Produktová specifikace
Model Cutstar
Environmentally Preferable
Obouruké/jednoruké Jednoruké
Jednorázové/znovupoužitelné opakovatelně použitelné
Materiál vložky Syntetická vlákna
Provedení PVC dot coating
Rozchod 13
Typ manžety Pletené zápěstí
Norma EN EN 388:2016 (3X42B)