Tisk…
Industrial gloves, Ultrane 551
Dodavatel: MAPA
34551030 34551031 34551036 34551037 34551038 34551039
MAPA34551030EA 7590 CZK
MAPA34551030 MAPA34551031 MAPA34551036 MAPA34551037 MAPA34551038 MAPA34551039
Industrial gloves, Ultrane 551
Rukavice
These seamless knitted gloves with PU foam coating on palm and fingertips provide excellent abrasion resistance.
  • Excellent tactile sensitivity due to reduced glove thickness
  • Absorption of perspiration ensured by the cellular structure of the PU coating
  • Ventilated back for additional wearing comfort
Typical applications include: Automotive/mechanical industry, sorting small parts, fitting small screws and fasteners, electronic component assembly (displays, LEDs, etc.), high-frequency antennae riveting, microprocessor handling, food industry, cosmeticsMore Product Information