Tisk…
NANOCOLOR® Photometric Standard Test Kit for Zinc
Dodavatel: MACHEREY-NAGEL
91895
MANA91895EA 5090 CZK
MANA91895
NANOCOLOR® Photometric Standard Test Kit for Zinc
Kity pro environmentální testy
Photometric test kit for the determination of zinc in water samples.
  • Ready-to-use reagents for up to 500 determinations
  • Excellent sensitivity and accuracy due to measurements in 50 mm cuvettes
  • Low costs per determination
  • Easy dilutions for analysis of higher concentrations
The test kit is evaluated in 10 or 50 mm rectangular cuvettes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II
Informace o dodávce: Rugged box with reagents for 250 tests.