Tisk…
QUANTOFIX Iron Sensitive 1
Dodavatel: MACHEREY-NAGEL
91359
MANA91359EA 464 CZK
MANA91359
QUANTOFIX Iron Sensitive 1
Kity pro environmentální testy
Semi-quantitative test strips for reliable determination of iron concentrations as low as 0.05 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity by a factor of nearly 10. Ideal for rapid and easy analysis of iron directly on-site. No maintenance or calibration is required.
  • Sensitive rapid test