Tisk…
Test strips, potassium
Dodavatel: MACHEREY-NAGEL
90727
MANA90727EA 1790 CZK
MANA90727
Test strips, potassium
Kity pro environmentální testy
The test paper allows the quick and easy detection of Potassium. Rubidium, Cesium and Thallium(I) give the same reaction. Sodium and heavy metal ions decrease the sensitivity.
  • Colour changes: Bright-yellow to orange
The leaflet describes a method to eliminate these interferences.