Tisk…

Cellulose MN 2100ff adsorbents for column chromatography (cellulose MN 2100 defatted)

Dodavatel: MACHEREY-NAGEL

815070.1
MANA815070.1EA 5090 CZK
MANA815070.1
Cellulose MN 2100ff adsorbents for column chromatography (cellulose MN 2100 defatted)
Cellulose

Native fibrous cellulose, purified grade (washed with different eluents) with average degree of polymerization 620–680.
Grade MN 2100ff is a defatted cellulose MN 2100 with a dichloromethane extract <0.02%.


  • For separation of hydrophilic substances
  • Fiber length (85%): 20 to 75 μm
  • Specific surface according to Blaine ~5500 cm²/g

Vzorec: (C₁₂H₂₂O₁₁)n
MDL Number: MFCD00081512
CAS číslo: 9004-34-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR