Tisk…
Capillary columns, OPTIMA® FFAP
Dodavatel: MACHEREY-NAGEL
726116.25 726116.30 726116.50 726116.60 726341.25 726341.30 726341.50 726341.60 726344.25 726344.30 726344.50 726345.30 726180.10 726346.25
MANA726116.25EA 18990 CZK
MANA726116.25 MANA726116.30 MANA726116.50 MANA726116.60 MANA726341.25 MANA726341.30 MANA726341.50 MANA726341.60 MANA726344.25 MANA726344.30 MANA726344.50 MANA726345.30 MANA726180.10 MANA726346.25
Capillary columns, OPTIMA® FFAP
Kolony chromatografické
Polar GC phase.
  • Ideal for fatty acid methyl esters (FAMEs), free carboxylic acids
  • Maximum temperature for isothermal operation 250 °C (0,53 mm ID columns: 220 °C)
  • Maximum temperature for short isotherms in a temperature program 260 °C (0,53 mm ID columns: 240 °C)
Polyethylene glycol 2-nitroterephthalate phase.
USP code: G35 / close equivalent to G25.