Tisk…
Glass microfibre filters without binder, grade MN 85/220 BF
Dodavatel: MACHEREY-NAGEL
408007 4080045 408014 4080025
MANA408007EA 4410 CZK
MANA408007 MANA4080045 MANA408014 MANA4080025
Glass microfibre filters without binder, grade MN 85/220 BF
Filtry Filtry ze skleněných mikrovláken
Binder free filter paper for difference weight analysis with very high loading capacity. This paper thick paper has a very good particle retention for fine particles.
  • Thermally stable up to 500 °C
  • Extremely high loading capacity
  • Binder-free