Tisk…
Glass microfiber filters with organic binder, grade MN 85/220
Dodavatel: MACHEREY-NAGEL
407024 407007 407018 4070025 4070037 4070045 407005 407009 407011 407012 407015 407027
MANA407024EA 13170 CZK
MANA407024 MANA407007 MANA407018 MANA4070025 MANA4070037 MANA4070045 MANA407005 MANA407009 MANA407011 MANA407012 MANA407015 MANA407027
Glass microfiber filters with organic binder, grade MN 85/220
Filtry Filtry ze skleněných mikrovláken
MN 85/220 is a thick glass fiber paper with high basis weight for the filtration of large volumes of liquids.
  • For large volumes of sample
  • Extemely high loading capacity
  • Good mechanical stability