Tisk…

L(+)-Arginin 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Dodavatel: Avantor Performance Materials

Synonyms: H-L-Arg-OH, (S)-2-Amino-5-guanidinopentanoic acid, Arginine, (S)-(+)-Arginine, L-Arginine, H-Arg-OH

MACR4926-06EA 0 CZK
MACR4926-06 MACR4926-07 MACR4926-09
L(+)-Arginin 98.5-101.0% USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L(+)-Arginin
Vzorec: C₆H₁₄N₄O₂
Bod varu: 368 °C
Bod tání: 238 °C
Hustota: 1,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS číslo: 74-79-3
EINECS: 200-811-1

Objednat

Výsledky specifikačního testu

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.5%
USP - Identification Passes Test
USP - Specific Rotation (A]25 AO (+) 26.3 - 27.7°
USP - Loss on Orying at 105 °C ≤0.5%
USP - Chloride (Cl) ≤0.05%
USP - Residue on Ignition ≤0.3%
USP - Sulfate (S04) ≤0.03%
USP - Iron (Fe) ≤0.003%
USP - Related Compounds - Individual Impurities ≤0.5%
USP - Related Compounds - Total Impurities ≤2.0%
EP/BP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation (A]2°AO (+) 25.5 - 28.5 °
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Chloride (Cl) ≤200ppm
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Orying ≤0.5%
EP/BP - Ash (sulfated) ≤0.1%
JP - Identification Passes Test
JP - 0ptical rotation, 20 °C (+) 26.9 - 27.9°
JP - pH (1:10) 10.5 - 12.0
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021%
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Iron (Fe) ≤10 ppm
JP - Related Substances Passes Test
JP - Loss on Orying at 105 °C ≤0.30%
JP - Residue after Ignition ≤0.1%
JP - Assay (C6Hl4N402)(dried basis) 98.5 - 101.0%
Endotoxin Concentration, IU/mg, For Information only
ChP - Specific Rotation @ 20 °C 26.9 - 27.9
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Alkalinity (pH) 10.5 - 12.0
ChP - Transmittance of solution 0.98
ChP - Chlorides ≤0.02%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - Proteins Passes Test
ChP - 0ther amino acids (TLC)(<=0.4%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.5%
ChP - Residue on ignition ≤0.1%
ChP - Iron ≤0.001%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Assay, dried basis 99.0 - 101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR