Tisk…

Guanidiniumchlorid ≥99.5%, Proteomics Grade

Dodavatel: VWR Chemicals
Varování

Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride

M110-500GEA 10080 CZK
M110-500G
Guanidiniumchlorid ≥99.5%, Proteomics Grade
Guanidiniumchlorid

Suited for purification of nucleic acids or polypeptides from cell sources and inhibition of both RNase and protease activity.

Vzorec: NH₂C(=NH)NH₂·HCl
Bod tání: 185 °C
Hustota: 1,354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS číslo: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Objednat

Výsledky specifikačního testu

Abs.@230 nm (6 M, Water) <0.15
Abs.@260 nm (6 M, Water) <0.03
Arsenic <0.0005%
DNase none detected
Iron <0.0005%
Lead <0.0005%
Melting Point 181 – 187 °C
Protease none detected
Purity >99.5%
RNase none
Solubility (6 M, water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR