Tisk…
Urine containers, with screw cap and spout
Dodavatel: LP ITALIANA
108039
LPIT108039EA 4740 CZK
LPIT108039
Urine containers, with screw cap and spout
Systémy pro klinickou diagnostiku Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
Container made of HDPE.
  • Good chemical resistance
  • Good temperature resistance