Tisk…
Test kits, ozone (kits and discs), CHECKIT®
Dodavatel: Lovibond
146275 147270 513170BT 513171BT 146270 147275
LOVI146275EA 825 CZK
LOVI146275 LOVI147270 LOVI513170BT LOVI513171BT LOVI146270 LOVI147275
Test kits, ozone (kits and discs), CHECKIT®
Kity pro environmentální testy
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
  • Enables the use of large path lengths
  • The mirror optics makes use of the view through the entire length of the cell
  • Easy operation and exact reagent dosing
  • Tablet reagents have minimum guaranteed shelf life of 5/10 years
The operating instructions provide a step-by-step explanation of conducting the water test to achieve reliable and accurate measurements in the minimum of time.
CHECKIT® disc contains a continuous colour scale which makes it possible to achieve an exact colour match between the colour standard and the sample. These discs are specially manufactured in selected materials to remain colour-stability over a long period and guarantee reliable, reproducible measurement results.
Informace o dodávce: Test kit includes CHECKIT® discs, cells, stirring rod and AQUALYTIC® tablet reagents (for 30 tests) for the desired test supplied in a sturdy plastic case.