Tisk…
Films for acrylamide gels, GelBond® PAG
Dodavatel: LONZA
54723 54727 54711 54729 54731 54733 54746
LONZ54723EA 20440 CZK
LONZ54723 LONZ54727 LONZ54711 LONZ54729 LONZ54731 LONZ54733 LONZ54746
Films for acrylamide gels, GelBond® PAG
Těsnění Filmy těsnící
GelBond® PAG film is a transparent, flexible polyester film designed to support polyacrylamide or MDE™ gels. Gels remain permanently attached to the film through electrophoresis and all subsequent fixing, staining, destaining and drying procedures.
  • Polyacrylamide gels retain their original dimensions during staining and after drying
  • Gels, particularly thin ones, are easier to handle during staining, destaining, and drying when supported
  • Gel orientation can be recorded directly on the GelBond® PAG film prior to casting