Tisk…
Contact Plates, Gamma-Irradiated, Individually Packed
Dodavatel: LIOFILCHEM
15339S 15323S 15327S 15340S 15380S 15360 15336S 15330 15330S 15333S 15341S 15345S 15365S
LIOF15339SEA 1740 CZK
LIOF15339S LIOF15323S LIOF15327S LIOF15340S LIOF15380S LIOF15360 LIOF15336S LIOF15330 LIOF15330S LIOF15333S LIOF15341S LIOF15345S LIOF15365S
Contact Plates, Gamma-Irradiated, Individually Packed
Média mikrobiologická Misky mikrobiologické kontaktní
Contact plates are used for monitoring surfaces, air, and personnel inside cleanrooms and sterile areas. The peelable blister pack ensures the highest quality and protection standards for the plates and reduces waste.
  • Individual plate packaging reduces waste by keeping unused plates valid until shelf life ends
  • Transparent packaging allows easy plate examination before opening
  • Drastically reduces the dehydration of culture media during the shelf life period
  • Shelf life up to 9 months
  • Storage at ambient temperature for most items in its range
  • Lowers contamination risks in controlled environments
Available as gamma-irradiated, triple-enveloped plates, suitable for restricted areas such as isolators and cleanrooms.
Certifikace:
In compliance with European Pharmacopoeia, United States Pharmacopeia, and Japanese Pharmacopoeia.