Tisk…
Contact Plates, Blister
Dodavatel: LIOFILCHEM
15325 15327 15328 15329 15331 15332 15362 15334 15365 15326 15336 15375 15376 15205 15203S 15204S 15389 15361
LIOF15325EA 1680 CZK
LIOF15325 LIOF15327 LIOF15328 LIOF15329 LIOF15331 LIOF15332 LIOF15362 LIOF15334 LIOF15365 LIOF15326 LIOF15336 LIOF15375 LIOF15376 LIOF15205 LIOF15203S LIOF15204S LIOF15389 LIOF15361
Contact Plates, Blister
Média mikrobiologická Misky mikrobiologické kontaktní
The peelable blister pack fulfills the highest quality and protection standards for the plate. The blister pack, produced fully automatically and in controlled environment, offers the following advantages compared to the pouch pack:
  • Full transparency of the package
  • No release particles during the film opening phase, critical in environment controlled by particle counters
  • Allows the usage of single plates, leaving the remaining plates protected in their pack
  • Drastically reduces the dehydration of culture medium during the shelf life period
  • Permits better product preservation and isolation during transportation and storage
  • Guarantees the product cannot be handled after the packaging and avoids contamination