Tisk…
Filter funnels
Dodavatel: Lenz Laborglas GmbH & CO.KG
07195428 07195379 07195328 07195334 07195340 07195358 07195434 07195440 07195458 07195258 07195128 07195134 07195140 07195158 07195170 07195179 07195228 07195234 07195240 07195270 07195279 07195370 07195470 07195479
LENZ07195428EA 1340 CZK
LENZ07195428 LENZ07195379 201-0594 LENZ07195334 LENZ07195340 LENZ07195358 LENZ07195434 LENZ07195440 LENZ07195458 LENZ07195258 LENZ07195128 LENZ07195134 LENZ07195140 LENZ07195158 LENZ07195170 LENZ07195179 LENZ07195228 LENZ07195234 LENZ07195240 LENZ07195270 LENZ07195279 LENZ07195370 LENZ07195470 LENZ07195479
Filter funnels
Nálevky Nálevky filtrační Nálevky k filtraci
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.