Tisk…
Reaction vessels, cylindrical, with flat flange and thermal jacket
Dodavatel: Lenz Laborglas GmbH & CO.KG
06011258 06011270 06011273 06011137 06011149 06011158 06011249 06011276 06011279 06011473 06011476 06011479 06011482 06011485 06011488
LENZ06011258EA 18110 CZK
LENZ06011258 LENZ06011270 201-0018 LENZ06011137 LENZ06011149 LENZ06011158 LENZ06011249 LENZ06011276 LENZ06011279 LENZ06011473 LENZ06011476 LENZ06011479 LENZ06011482 LENZ06011485 LENZ06011488
Reaction vessels, cylindrical, with flat flange and thermal jacket
Nádoby reakční Nádoby reakční, cylindrický tvar
DURAN®, borosilicate glass, clear, with flat bottom.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and flat laboratory flange (LF) with groove. With two branches KF 15.