Tisk…
Apparatus - Antonacopoulos
Dodavatel: Lenz Laborglas GmbH & CO.KG
05434000 05434001 05434002 05434004 05434005
LENZ05434000EA 20470 CZK
LENZ05434000 LENZ05434001 LENZ05434002 LENZ05434004 LENZ05434005
Apparatus - Antonacopoulos
Komponenty pro destilaci Destilační soupravy
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes