Tisk…
Flasks, round bottom with four necks and standard ground joints, side necks angled (7°)
Dodavatel: Lenz Laborglas GmbH & CO.KG
03347358 03347458 03347449 03347470 03347349
LENZ03347358EA 3410 CZK
LENZ03347358 LENZ03347458 LENZ03347449 LENZ03347470 LENZ03347349
Flasks, round bottom with four necks and standard ground joints, side necks angled (7°)
Baňky Baňky s kulatým dnem
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certifikace: DIN 12392