Tisk…
Dry evaporation systems, N₂/48, RapidVap®
Dodavatel: LABCONCO
7910013 7482100 7482200
LBCP7910013EA 604770 CZK
LBCP7910013 LBCP7482100 LBCP7482200
Dry evaporation systems, N₂/48, RapidVap®
Odparky Centrifugační odpařování
This system uses nitrogen blow down, vortex motion and heat to speed evaporation of up to 48 small samples or eight 450 milliliter samples at once.
  • Nitrogen blow down reduces partial pressure directly over the liquid to speed up the evaporation
  • Nitrogen may be introduced to 2, 4, 6 or 8 clusters of 6 sample tubes
  • Microprocessor-controlled 1000-watt dry block heating system supplies a controlled amount of heat up to 100 °C
  • Two-way RS232 link to allow remote control of functions from a user-supplied personal computer
The system may be used to prepareMore Product Information