Tisk…
Glove boxes, controlled atmosphere, Protector®
Dodavatel: LABCONCO
5060133 5060135 5080135 5060131 5060137 5080137 5080131 5080133
LBCP5060133EA 2185670 CZK
LBCP5060133 LBCP5060135 LBCP5080135 LBCP5060131 LBCP5060137 LBCP5080137 LBCP5080131 LBCP5080133
Glove boxes, controlled atmosphere, Protector®
Gloveboxy
These glove boxes provides a leak-tight environment to perform work with contamination-sensitive materials under vacuum or in an inert environment.
  • Performance tested to maintain oxygen levels as low as 0,2 ppm and moisture levels as low as 1 ppm
  • Epoxy-coated aluminum double-grooved glove ports with neoprene gaskets
  • Space-saving inner and outer transfer chamber doors that pivot upward, are counterbalanced and equipped with quick-latches
  • Interior right-hand side wall-mounted electrical duplex receptacle with 10 amp maximum (230 volt models) circuit load
More Product Information