Tisk…
Vertical laminar air flow cabinets, Purifier®
Dodavatel: LABCONCO
3970324 3970421
LBCP3970324EA 0 CZK
LBCP3970324 LBCP3970421
Vertical laminar air flow cabinets, Purifier®
Digestoře Boxy laminární
Purifier Vertical Clean Benches direct HEPA-filtered air downward over the work area to provide ISO Class 5 (formerly Class 100) conditions and protect your work from particulate contamination. Applications for clean benches include plant tissue culture, media plate preparation, electronics inspection, medical device assembly and pharmacy drug preparation.
  • Nominal downflow velocity of 45 to 65 fpm with aerodynamic air foil
  • Disposable prefilter and variable speed built-in blower with motorized impeller and solid state control rated for 10 amps
  • ISO Class 5
More Product Information