Tisk…
XPert™ Nano Ventilated Enclosures
Dodavatel: LABCONCO
3887462 3887221 3887421 3887521 3888562 3887662 3887562 3887262
LBCP3887462EA 0 CZK
LBCP3887462 LBCP3887221 LBCP3887421 LBCP3887521 LBCP3888562 LBCP3887662 LBCP3887562 LBCP3887262
XPert™ Nano Ventilated Enclosures
Digestoře Odtahy
In nanoparticle manipulation and dry powder chemical handling, enclosures keep hazardous powders and particulates contained. Room air is pulled into the enclosure from the front, flows to the baffle, and finally passes through a ULPA exhaust filter (99,999% efficient on particles 0,12 μm in size) before returning to the laboratory or cleanroom.
  • Built-in ioniser neutralises static charge on interior surfaces by emitting ions into the airstream, which reduces weighing errors and the attraction of powders to surfaces
  • ULPA filter is top-accessible for leak