Tisk…
Jerricans, narrow neck, HDPE
Dodavatel: LAMAPLAST
3952 3951 3950 3949 3948
LAMA3952EA 917 CZK
LAMA3952 LAMA3951 LAMA3950 LAMA3949 LAMA3948
Jerricans, narrow neck, HDPE
Láhve Kanystry
HDPE, translucent
  • Supplied with pouring spout (except for 5 L jerrican)
  • Spout is long enough for accurate pouring of liquids
  • With tethered sealing cap that cannot be lost
Universal canister for industry, trade and laboratories. Ideal for packaging, storage and transportation.