Tisk…
Polyethylene Heavy-Duty Graduated Buckets
Dodavatel: LAMAPLAST
161 162 163 165 164
LAMA161EA 347 CZK
LAMA161 LAMA162 LAMA163 LAMA165 LAMA164
Polyethylene Heavy-Duty Graduated Buckets
Kbelíky
Made with low density polyethylene (LDPE), in white or black colour options.
  • Conical shape, litre graduations
  • With ergonomic handle
  • Food contact material approved
Buckets feature a flat bottom design to ensure total emptying and allow for easy cleaning.
Certifikace: EU 10/2011.