Tisk…

N,N-Dimethylformamid

Dodavatel: Lach-ner S.R.O.
Nebezpečí

Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF

H33H11X
LACHH33H11XEA 0 CZK
LACHH33H11X
N,N-Dimethylformamid
N,N-Dimethylformamid
Vzorec: HCON(CH₃)₂
Bod varu: 153 °C (1013 hPa)
Bod tání: –61 °C
Hustota: 0,949 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS číslo: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR