Tisk…

Trilithium citrate tetrahydrate

Dodavatel: Lach-ner S.R.O.
Varování

Synonyms: Trilithium 2-hydroxypropane-1,2,3-tricarboxylate tetrahydrate, Lithium citrate tribasic tetrahydrate, Citric acid trilithium salt tetrahydrate

30094
LACH30094EA 0 CZK
LACH30094
Trilithium citrate tetrahydrate
Trilithium citrate tetrahydrate
Vzorec: C₆H₅Li₃O₇·4H₂O
MW: 281.99
MDL Number: MFCD00150030
CAS číslo: 6080-58-6
EINECS: 213-045-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR